Toluene-p-sulfonic anhydride 95%

Supplier: Thermo Scientific Chemicals
Danger

367070050 367070250 367071000
TS367070050EA 55 USD
TS367070050 TS367070250 TS367071000
Toluene-p-sulfonic anhydride 95%
Toluene-p-sulphonic anhydride
Formula: C₁₄H₁₄O₅S₂
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00008548
CAS Number: 4124-41-8
EINECS: 223-926-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white to beige to grey Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration by coupling ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR