2-(2-Ethoxyethoxy)ethyl acetate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethylene glycol monoethyl ether acetate
257420010
257420025
TS257420010EA
76
USD
TS257420010
TS257420025
2-(2-Ethoxyethoxy)ethyl acetate 99%
2-(2-Ethoxyethoxy)ethyl acetate
Formula:
C₈H₁₆O₄ MW: 176.21 g/mol Boiling Pt: 210…220 °C (760 mmHg) Melting Pt: –25 °C Density: 1.01 g/cm³ Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041928 CAS Number: 112-15-2 EINECS: 203-940-1 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4190 to 1.4230 (20°C, 589 nm) |
Water | ≤0.1 % (K.F.) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...