Methyl docosanoate 99% analytical standard for GC

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl behenate, Docosanoic acid methyl ester, Behenic acid methyl ester

226810010 226810050
TS226810010EA 61 USD
TS226810010 TS226810050
Methyl docosanoate 99% analytical standard for GC
Methyl docosanoate
Formula: C₂₃H₄₆O₂
MW: 354.62 g/mol
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00009347
CAS Number: 929-77-1

Order Now

Specification Test Results

Appearance White Flakes
Infrared spectrum Conforms
Melting point 53 to 56 °C
Solubility (5 % in Hexane) Clear colorless solution
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR