Erbium(III) perchlorate 40% (w/w) in water

Supplier: Thermo Scientific Chemicals
Danger

428290050 428290250
TS428290050EA 65 USD
TS428290050 TS428290250
Erbium(III) perchlorate 40% (w/w) in water
Erbium(III) perchlorate
Formula: Cl₃ErO₁₂
MW: 465.61
Density: 1.44 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00016073
CAS Number: 14017-55-1
UN: 3211
ADR: 5.1 II

Order Now

Specification Test Results

Appearance Clear pink to purple Liquid
Titration Complexometric 14 to 15 % (Er)
Thiosulphate (S2O3) ≤5000 ppm
Sulfite (as SO2) ≤2000 ppm
Total nitrogen (as N) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR