Cesium hydroxide 50% in water 99.9% (trace metals basis) for analysis

Supplier: Thermo Scientific Chemicals
Danger

213601000 213605000
TS213601000EA 121 USD
TS213601000 TS213605000
Cesium hydroxide 50% in water 99.9% (trace metals basis) for analysis
Cesium hydroxide
Formula: CsOH
MDL Number: MFCD00010964
CAS Number: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Additional info Trace analysis values are on dry salt basis
Titration with HCl 48 to 52 % (As CsOH)
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤25 ppm
Aluminium (Al) ≤5 ppm
Barium (Ba) ≤15 ppm
Calcium (Ca) ≤2 ppm
Chromium (Cr) ≤1 ppm
Iron (Fe) ≤1 ppm
Lithium (Li) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤1 ppm
Potassium (K) ≤200 ppm
Rubidium (Rb) ≤10 ppm
Sodium (Na) ≤100 ppm
Strontium (Sr) ≤5 ppm
Phosphorus pentoxide (P2O5) ≤5 ppm
Silicon dioxide (SiO2) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR