p-Toluenesulfonic acid 12% in acetic acid, pure

Supplier: Thermo Scientific Chemicals
Danger

421222500 421220010
TS421222500EA 174 USD
TS421222500 TS421220010
p-Toluenesulfonic acid 12% in acetic acid, pure
p-Toluenesulphonic acid
Formula: C₇H₈O₃S
Flash Pt: 41 °C
CAS Number: 104-15-4
EINECS: 203-180-0
UN: 2920

Order Now

Specification Test Results

Appearance Clear colorless to light yellow to pink Solution
Titration with NaOH 11 to 13 %
Melting point 84 to 88 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR