1-Butyl-3-methylimidazoliumtetrafluoroborate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium tetrafluoroborate, 1-n-Butyl-3-methylimidazolium tetrafluoroborate, [BMIM] [BF4]

354210050 354210250 354211000
TS354210050EA 47 USD
TS354210050 TS354210250 TS354211000
1-Butyl-3-methylimidazoliumtetrafluoroborate 98+%
1-Butyl-3-methylimidazoliumtetrafluoroborate
Formula: C₈H₁₅BF₄N₂
Melting Pt: –71 °C
Density: 1.2077 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD03095449
CAS Number: 174501-65-6
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow to orange Viscous liquid
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR