1-Butyl-3-methylimidazolium chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium chloride

354090050 354090250 354091000 354095000
TS354090050EA 42 USD
TS354090050 TS354090250 TS354091000 TS354095000
1-Butyl-3-methylimidazolium chloride 98+%
1-Butyl-3-methylimidazolium chloride
Formula: C₈H₁₅ClN₂
MW: 174.67 g/mol
Melting Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD03095425
CAS Number: 79917-90-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Cream to yellow to orange Powder or crystals or chunks or fused solid
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR