Tetraethylammonium perchlorate 0.2M in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

379801000 379805000
TS379801000EA 30 USD
TS379801000 TS379805000
Tetraethylammonium perchlorate 0.2M in aqueous solution
Tetraethylammonium perchlorate
Formula: C₈H₂₀ClNO₄
MW: 229.7 g/mol
Melting Pt: > 300 °C
Density: 1.0016 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00041977
CAS Number: 2567-83-1
UN: 3139
ADR: 5.1 II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.2 M Exact strength on the certificate of analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR