Barium thiosulfate, pure

Supplier: Thermo Scientific Chemicals
Warning

453811000 453815000
TS453811000EA 35 USD
TS453811000 TS453815000
Barium thiosulfate, pure
Barium thiosulphate
Formula: BaO₃S₂
Storage Temperature: Ambient
MDL Number: MFCD00040661
CAS Number: 35112-53-9
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Powder
Assay 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR