Methyl palmitate 99% analytical standard for GC

Supplier: Thermo Scientific Chemicals

Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

460390010 460390050
TS460390010EA 37 USD
TS460390010 TS460390050
Methyl palmitate 99% analytical standard for GC
Methyl palmitate
Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 338 °C (760 mmHg)
Melting Pt: 28 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008994
CAS Number: 112-39-0

Order Now

Specification Test Results

Appearance White Low melting solid
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR