Trichloroacetyl isocyanate for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

202900020 202900100 202900500
TS202900020EA 47 USD
TS202900020 TS202900100 TS202900500
Trichloroacetyl isocyanate for NMR spectroscopy
Trichloroacetyl isocyanate
Formula: C₃Cl₃NO₂
MW: 188.4 g/mol
Boiling Pt: 80…85 °C (20 mmHg)
Density: 1.59 g/cm³
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002033
CAS Number: 3019-71-4
EINECS: 221-165-7
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4799 to 1.4819 (20°C, 589 nm)
GC ≥97 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR