Tetrabutylammonium fluoride 1 M in THF

Supplier: Thermo Scientific Chemicals
Danger

201950025 201950050 201951000 201955000
TS201950025EA 774 USD
TS201950025 TS201950050 TS201951000 TS201955000
Tetrabutylammonium fluoride 1 M in THF
Tetrabutylammonium fluoride
Formula: C₁₆H₃₆FN
MW: 261.47
Density: 0.887 g/cm³
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011747
CAS Number: 429-41-4
EINECS: 207-057-2
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear yellow to amber to light green to brown Solution
Water ≤8 %
Titration with HClO4 0.90 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR