Starch, soluble 1% (w/w) in aqueous solution stabilised, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Starch, Vitex® iodine indicator, Amylodextrin

Stabilization: 100 ppb Stabilised with mercury (II) iodide

380641000 380645000
TS380641000EA 47 USD
TS380641000 TS380645000
Starch, soluble 1% (w/w) in aqueous solution stabilised, pure
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
Merck Index: 13,08878

Order Now

Specification Test Results

Appearance Clear to slightly hazy solution
Sensitivity Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR