Molecular sieve A4 (0.4 nm, 4 Å) 10 --> 18 mesh

Supplier: Thermo Scientific Chemicals

264755000 264750050
TS264755000EA 66 USD
TS264755000 TS264750050
Molecular sieve A4 (0.4 nm, 4 Å) 10 --> 18 mesh
Molecular sieve A4 (0.4 nm, 4 Å)
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now

Specification Test Results

Appearance Beige Granules
Bulk density 680 to 780 g/l
Residual water ≤1.5 % (50°C, 2 h)
Water adsorption ≥20 % (50% rH, 20°C, 24 h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR