Formamide 99% for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Carbamaldehyde

327230025 327235000
TS327230025EA 797 USD
TS327230025 TS327235000
Formamide 99% for molecular biology DNase-, RNase-, protease-free
Formamide
Formula: HCONH₂
Boiling Pt: 210 °C (760 mmHg)
Melting Pt: 2…3 °C
Density: 1.133 g/cm³
Flash Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0

Order Now

Specification Test Results

Appearance Clear liquid
Solubility (50 % in water) Complete
GC ≥99 %
UV at 280 nm A: ≤0.02
Color scale ≤5 APHA
pH 7 to 9 (1 M in 0.1 M KCl)
Dnase,Rnase,Protease activity None detected
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.5 ppm
Zinc (Zn) ≤0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR