Isobutanol 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

412650025 412655000
TS412650025EA 189 USD
TS412650025 TS412655000
Isobutanol 99+% ACS
Isobutanol
Formula: (CH₃)₂CHCH₂OH
Boiling Pt: 107.9 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0.8 g/cm³
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3945 to 1.3975 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
Solubility (in water) Passes test
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Carbonyl compounds ≤0.02 % 2-Butanone
Titrable acid ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR