Diisopropyl ether stabilized for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
390660010
390660025
TS390660010EA
95
USD
TS390660010
TS390660025
Diisopropyl ether stabilized for analysis
Diisopropyl ether
Formula:
(CH₃)₂CHOCH(CH₃)₂ Boiling Pt: 68 °C (760 mmHg) Melting Pt: –85.5 °C Density: 0.72 g/cm³ Flash Pt: –29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008880 CAS Number: 108-20-3 EINECS: 203-560-6 UN: 1159 ADR: 3,II |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Water | ≤0.1 % |
GC | ≥99.0 % |
Color scale | ≤25 APHA |
Stabilizer | Approx. 50 ppm BHT |
Non-volatile matter | ≤0.005 % |
Acidity | ≤0.0004 meq/g |
Aluminium (Al) | ≤0.5 ppm |
Barium (Ba) | ≤0.1 ppm |
Calcium (Ca) | ≤0.5 ppm |
Cobalt (Co) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Manganese (Mn) | ≤0.02 ppm |
Nickel (Ni) | ≤0.02 ppm |
Tin (Sn) | ≤0.1 ppm |
Zinc (Zn) | ≤0.1 ppm |
Peroxides | ≤0.005 % (as H2O2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...