Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

397210010 397211000
TS397210010EA 243 USD
TS397210010 TS397211000
Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal™
Bis(2-methoxyethyl)ether
Formula: C₆H₁₄O₃
Boiling Pt: 162 °C (760 mmHg)
Melting Pt: –64 °C
Density: 0.937 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00008503
CAS Number: 111-96-6
UN: 3271
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR