(S)-(+)-2-Butanol 99%

Supplier: Thermo Scientific Chemicals
Warning

218030010 218030050
TS218030010EA 65 USD
TS218030010 TS218030050
(S)-(+)-2-Butanol 99%
(S)-(+)-2-Butanol
Formula: C₄H₁₀O
Boiling Pt: 99 °C (1013 hPa)
Melting Pt: –114 °C
Density: 0.8 g/cm³
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00064281
CAS Number: 4221-99-2
EINECS: 224-168-1
UN: 1120
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3980 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation +12° to +14° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR