Benzyl alcohol ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
447720010
447720025
447721000
TS447720010EA
224
USD
TS447720010
TS447720025
TS447721000
Benzyl alcohol ACS
Benzyl alcohol
Formula:
C₆H₅CH₂OH Boiling Pt: 205 °C (760 mmHg) Melting Pt: –15 °C Density: 1.044 g/cm³ Flash Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 |
Specification Test Results
Appearance | Clear colorless Liquid |
GC | ≤0.01 % Benzaldehyde |
Color scale | ≤20 APHA |
Ash | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...