Dimethylamine 2 mol/l in methanol, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

370261000 370268000
TS370261000EA 81 USD
TS370261000 TS370268000
Dimethylamine 2 mol/l in methanol, AcroSeal®
Dimethylamine
Formula: C₂H₇N
MW: 45.08 g/mol
Density: 0.775 g/cm³
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Strength 1.9 to 2.2 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR