Lithium azide 20% in water, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
335360250
335361000
TS335360250EA
129
USD
TS335360250
TS335361000
Lithium azide 20% in water, pure
Lithium azide
Formula:
LiN₃ Storage Temperature: Ambient |
MDL Number:
MFCD00041956 CAS Number: 19597-69-4 EINECS: 243-177-1 UN: 3287 ADR: 6.1,I |
Specification Test Results
Appearance | Clear colorless Solution |
Titration Argentometric | 17 to 23 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...