Boron tribromide 1 M in heptane, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

378191000 378198000
TS378191000EA 111 USD
TS378191000 TS378198000
Boron tribromide 1 M in heptane, AcroSeal®
Boron tribromide
Formula: BBr₃
Boiling Pt: 90 °C
Density: 0.869 g/cm³
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00011312
CAS Number: 10294-33-4
EINECS: 233-657-9
UN: 3384
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear yellow to amber Solution
Strength 0.9 to 1.1 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR