Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture, anhydrous 99%, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Decalin, Bicyclo[4.4.0]decane, Decahydronaphthalene

406170010 406171000
TS406170010EA 150 USD
TS406170010 TS406171000
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture, anhydrous 99%, AcroSeal®
Decahydronaphthalene (mixture of cis and trans isomers)
Formula: C₁₀H₁₈
MW: 138.25 g/mol
Boiling Pt: 187 °C (760 mmHg)
Melting Pt: –31 °C
Density: 0.89 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004130
CAS Number: 91-17-8
EINECS: 202-046-9
UN: 1147
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.5 % (mixture of cis and trans)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR