Lithiumbis(trimethylsilyl)amide 1.0 mol/l in tert-butyl methyl ether, AcroSeal®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lithiumbis(triméthylsilyl)amide, HMDS-Li, N-Lithiohexamethyldisilazane, HMDS lithium salt
345671000
345678000
TS345671000EA
189
USD
TS345671000
TS345678000
Lithiumbis(trimethylsilyl)amide 1.0 mol/l in tert-butyl methyl ether, AcroSeal®
Lithiumbis(trimethylsilyl)amide
Formula:
C₆H₁₈LiNSi₂ Boiling Pt: 55…56 °C (760 mmHg) Density: 0.8 g/cm³ Flash Pt: –28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008261 CAS Number: 4039-32-1 EINECS: 223-725-6 UN: 2924 ADR: 3,II |
Specification Test Results
Appearance | Clear to hazy yellow to brown Solution |
Titration Active base | 21 to 25 % (as LiNSi) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...