1.0 M in water

Supplier: Thermo Scientific Chemicals

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

397080010 397080025
TS397080010EA 55 USD
TS397080010 TS397080025
1.0 M in water
di-Potassium hydrogen phosphate
Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
Merck Index: 13,07743

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HClO4 1.0 M Exact strength on the certificate of analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR