(R)-(-)-2-Butanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol

217230010 217230050 217230250
TS217230010EA 69 USD
TS217230010 TS217230050 TS217230250
(R)-(-)-2-Butanol 99%
(R)-(-)-2-Butanol
Formula: C₄H₁₀O
Boiling Pt: 99…100 °C (760 mmHg)
Melting Pt: –114 °C
Density: 0.8 g/cm³
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00064280
CAS Number: 14898-79-4
EINECS: 238-967-8
UN: 1120
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3980 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -12° to -14° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR