Potassium hydroxide ca. 85%, pellets ACS

Supplier: Thermo Scientific Chemicals
Danger

424140025 424140050 424140250 424140251 424145000
TS424140025EA 158 USD
TS424140025 TS424140050 TS424140250 TS424140251 TS424145000
Potassium hydroxide ca. 85%, pellets ACS
Potassium hydroxide
Formula: KOH
Boiling Pt: 1320 °C (760 mmHg)
Melting Pt: 360 °C
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
REACH: 01-2119487136-33

Order Now

Specification Test Results

Appearance White Pellets
Titration with HCl ≥85 % (KOH)
Chloride (Cl) ≤0.01 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.003 %
Potassium carbonate ≤2.0 % (K2CO3)
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.002 %
Nickel (Ni) ≤0.001 %
Sodium (Na) ≤0.05 %
Heavy metals (ICP-OES) ≤0.001 %
Nitrogen compounds (as N) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR