Maleic anhydride, briquettes

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,5-Furanedione

465660010 465660050 465660250 465660251
TS465660010EA 35 USD
TS465660010 TS465660050 TS465660250 TS465660251
Maleic anhydride, briquettes
Maleic anhydride

  • Organic reagent

Formula: C₄H₂O₃
MW: 98.06 g/mol
Boiling Pt: 200 °C (760 mmHg)
Melting Pt: 52…55 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00005518
CAS Number: 108-31-6
UN: 2215
ADR: 8,III

Order Now

Specification Test Results

Appearance White Briquettes
Infrared spectrum Conforms
Melting point 50 to 56 °C
Free acid ≤0.5 (as maleic acid)
Titration with NaOH 98.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR