1,2-Dimethoxyethane, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethyl glycol, Ethylene glycol dimethyl ether, DME
442980010
442981000
TS442980010EA
282
USD
TS442980010
TS442981000
1,2-Dimethoxyethane, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
1,2-Dimethoxyethane
Formula:
C₄H₁₀O₂ MW: 90.12 g/mol Boiling Pt: 85 Melting Pt: -69 Density: 0.867 g/cm³ Flash Pt: –6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008502 CAS Number: 110-71-4 UN: 2252 ADR: 3 II Merck Index: 13,03251 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % |
GC | ≥99.4 % |
Stabilizer | Approx. 100 ppm BHT |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...