tert-Butyl methyl ether ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether

447470010 447470025
TS447470010EA 65 USD
TS447470010 TS447470025
tert-Butyl methyl ether ACS
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
Boiling Pt: 54…56 °C (760 mmHg)
Melting Pt: –110 °C
Density: 0.7404 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
UN: 2398
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 %
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Peroxides ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR