Methanesulfonic anhydride 97%

Supplier: Thermo Scientific Chemicals
Danger

348760050 348760250 348761000 348765000
TS348760050EA 27 USD
TS348760050 TS348760250 TS348761000 TS348765000
Methanesulfonic anhydride 97%
Formula: C₂H₆O₅S₂
Boiling Pt: 125 °C (4 mmHg)
Melting Pt: 69…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00007556
CAS Number: 7143-01-3
EINECS: 230-442-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to grey to light brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration by coupling ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR