Dimethyl sulphoxide 99.8+% for HPLC
Supplier: Thermo Scientific Chemicals
Synonyms:
Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
397600010
397600025
TS397600010EA
152
USD
TS397600010
TS397600025
Dimethyl sulphoxide 99.8+% for HPLC
Dimethyl sulphoxide
Formula:
(CH₃)₂SO Boiling Pt: 189 °C (760 mmHg) Melting Pt: 18.4 °C Density: 1.1 g/cm³ Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.1 % |
GC | ≥99.80 % |
UV | at 400 nm A: ≤0.009 |
Residue after evaporation | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...