Iodine monochloride 1 M in dichloromethane
Supplier: Thermo Scientific Chemicals
|
Danger
|
370741000
370748000
TS370741000EA
73
USD
TS370741000
TS370748000
Iodine monochloride 1 M in dichloromethane
Iodine monochloride
Formula:
ClI Storage Temperature: Ambient |
MDL Number:
MFCD00011354 CAS Number: 7790-99-0 EINECS: 232-236-7 UN: 2922 ADR: 8,II |
Specification Test Results
Appearance | Orange to red to brown Solution |
Titration with Na2S2O3 | 0.95 to 1.05 M |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...