Titanium(IV) chloride 1 M in dichloromethane, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Titanium tetrachloride

377741000 377748000
TS377741000EA 74 USD
TS377741000 TS377748000
Titanium(IV) chloride 1 M in dichloromethane, AcroSeal®
Titanium(IV) chloride
Formula: Cl₄Ti
MW: 189.69
Boiling Pt: 136
Melting Pt: 136.2
Density: 1.72
Storage Temperature: Ambient
MDL Number: MFCD00011267
CAS Number: 7550-45-0
EINECS: 231-441-9
UN: 2922
ADR: 6.1 II
Merck Index: 13,09555

Order Now

Specification Test Results

Appearance Light yellow to dark brown Solution
Titration Argentometric 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR