Osmium tetraoxide 2.5% (w/w) in tert-butanol stabilized

Supplier: Thermo Scientific Chemicals
Danger

197450010 197450050 197450250
TS197450010EA 50 USD
TS197450010 TS197450050 TS197450250
Osmium tetraoxide 2.5% (w/w) in tert-butanol stabilized
Osmium tetraoxide
Formula: O₄Os
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011150
CAS Number: 20816-12-0
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance White to yellow Fused mass or solution
Stabilizer tert-Butyl hydroperoxide
Strength ≥2.5 wt%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR