Phosphorus(III) bromide 1 mol/l in dichloromethane, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

382061000 382068000
TS382061000EA 76 USD
TS382061000 TS382068000
Phosphorus(III) bromide 1 mol/l in dichloromethane, AcroSeal®
Phosphorus(III) bromide
Formula: PBr₃
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
UN: 2922

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution
Titration Argentometric 0.9 to 1.1 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR