Orthophosphoric acid 85% in water, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
295700010
295700025
295700100
TS295700010EA
90
USD
TS295700010
TS295700025
TS295700100
Orthophosphoric acid 85% in water, Extra Pure
Orthophosphoric acid
Formula:
H₃PO₄ Melting Pt: 21 °C Density: 1.68 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 EINECS: 231-633-2 UN: 1805 ADR: 8,III |
Specification Test Results
Appearance | Clear liquid |
Color scale | ≤35 APHA |
Titration with NaOH | 84.5 to 86.0 % |
Chloride (Cl) | ≤10 ppm |
Arsenic (As) | ≤2 ppm |
Iron (Fe) | ≤20 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...