Water, extra pure, deionized
Supplier: Thermo Scientific Chemicals
Synonyms:
Hydrogen oxide
345470050
345470250
TS345470050EA
57
USD
TS345470050
TS345470250
Water, extra pure, deionized
Water
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 Melting Pt: 0 Density: 1 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Specification Test Results
Appearance | Clear colorless Liquid |
Residue after evaporation | ≤25 ppm |
Conductivity | ≤5.0 µS/cm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...