Sodium borohydride 12% in aqueous solution, VenPure™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

448510025 448511000 448515000
TS448510025EA 205 USD
TS448510025 TS448511000 TS448515000
Sodium borohydride 12% in aqueous solution, VenPure™
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Boiling Pt: 130…135 °C (760 mmHg)
Melting Pt: 400
Density: 1.4 g/cm³
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 3320
ADR: 8 II
Merck Index: 13,08664

Order Now

Specification Test Results

Appearance Clear liquid
Color scale ≤50 APHA
Titration with HCl 38.0 to 42.0 % w/w (NaOH)
Titration with Na2S2O3 11.85 to 12.15 % w/w (NaBH4)
Iron (Fe) ≤10 ppm
Methanol ≤2000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR