Lithium aluminum hydride 4.0 M in diethyl ether, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

199511000 199518000
TS199511000EA 78 USD
TS199511000 TS199518000
Lithium aluminum hydride 4.0 M in diethyl ether, AcroSeal®
Lithium aluminum hydride
Formula: H₄AlLi
Flash Pt: –017 °C
Storage Temperature: Ambient
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1411
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Colorless Solution (May develop some turbidity or precipitate)
Titration Iodometric 18.0 to 22.0 % (LiAlH4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR