Lithium aluminum hydride 2.4 mol/l in THF, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate

377321000 377328000
TS377321000EA 95 USD
TS377321000 TS377328000
Lithium aluminum hydride 2.4 mol/l in THF, AcroSeal®
Lithium aluminum hydride
Formula: H₄AlLi
MW: 37.95 g/mol
Melting Pt: 125
Flash Pt: –17 °C
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1411
ADR: 4.3 I
Merck Index: 13,00348

Order Now

Specification Test Results

Appearance Colorless to grey Solution (May develop some turbidity or precipitate)
Strength 9.5 to 10.5 % (as LiAlH4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR