Lithium aluminum hydride 1 M in THF, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

199491000 199498000
TS199491000EA 60 USD
TS199491000 TS199498000
Lithium aluminum hydride 1 M in THF, AcroSeal®
Lithium aluminum hydride
Formula: H₄AlLi
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1411
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Turbid grey Solution
Titration Iodometric 3.9 to 4.5 % (as LiAlH4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR