1,4-Dioxane 99.5% stabilised for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

270490025 270490010
TS270490025EA 217 USD
TS270490025 TS270490010
1,4-Dioxane 99.5% stabilised for analysis
1,4-Dioxane
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.1
Melting Pt: 11.8
Density: 1.034 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3 II
Merck Index: 13,03330

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 %
GC ≥99.4 %
Color scale ≤10 APHA
Stabilizer 5 to 30 ppm BHT
Cadmium (Cd) ≤0.1 ppm
Chromium (Cr) ≤0.1 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.1 ppm
Free acid (CH3COOH) ≤0.001 %
Peroxides ≤0.005 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR