Cyclohexanone 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pimelic Ketone
406090010
406090025
406092500
TS406090010EA
73
USD
TS406090010
TS406090025
TS406092500
Cyclohexanone 99+% ACS
Cyclohexanone
Formula:
C₆H₁₀(=O) Boiling Pt: 155 °C (760 mmHg) Melting Pt: –47 °C Density: 0.947 g/cm³ Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001625 CAS Number: 108-94-1 UN: 1915 ADR: 3,III |
Specification Test Results
Appearance | Clear viscous liquid |
Water | ≤0.05 % |
GC | ≥99.0 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...