Nitrobenzene 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

415780025 415780250 415785000
TS415780025EA 311 USD
TS415780025 TS415780250 TS415785000
Nitrobenzene 99+% ACS
Nitrobenzene
Formula: C₆H₅NO₂
Boiling Pt: 210.8 °C (1013 hPa)
Melting Pt: 5…6 °C
Density: 1.205 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00007043
CAS Number: 98-95-3
EINECS: 202-716-0
UN: 1662
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5540 (20°C, 589 nm)
GC ≥99.0 %
Residue after evaporation ≤0.005 %
Chloride (Cl) ≤5 ppm
Water soluble titr. acid ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR