Chlorobenzene 99+% for spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

222110025 222115000
TS222110025EA 268 USD
TS222110025 TS222115000
Chlorobenzene 99+% for spectroscopy
Chlorobenzene
Formula: C₆H₅Cl
Boiling Pt: 131 °C (760 mmHg)
Melting Pt: –45 °C
Density: 1.1075 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45

Order Now

Specification Test Results

Appearance Clear liquid
GC ≥99.0 %
UV at 360 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR