2-Methylbutane HPLC
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
iso-Pentane
427730010
427730025
TS427730010EA
216
USD
TS427730010
TS427730025
2-Methylbutane HPLC
2-Methylbutane
Formula:
CH₃CH₂CH(CH₃)₂ MW: 72.15 g/mol Boiling Pt: 28 Melting Pt: -160 Density: 0.62 g/cm³ Flash Pt: –51 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009338 CAS Number: 78-78-4 UN: 1265 ADR: 3 I |
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.02 % (Coulometric) |
GC | ≥99.0 % |
UV | at 195 nm A: ≤1.0 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...