2-Methylbutane, dry_extra 99+%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
iso-Pentane
397220010
397221000
TS397220010EA
194
USD
TS397220010
TS397221000
2-Methylbutane, dry_extra 99+%, AcroSeal™
2-Methylbutane
Formula:
CH₃CH₂CH(CH₃)₂ MW: 72.15 g/mol Boiling Pt: 28 Melting Pt: -160 Density: 0.62 g/cm³ Flash Pt: –51 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009338 CAS Number: 78-78-4 UN: 1265 ADR: 3 I |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % |
GC | ≥99.0 % |
Residue after evaporation | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...